Product Details

SNP ID
rs143155041
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:95089482 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTCAAGAGACAGGCCAGCCCTCCTG[A/G]GCAGGTAGGGGAGCTGCCCAAGAAT
Phenotype
MIM: 611972
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MRPS5 PubMed Links

Gene Details

Gene
MRPS5
Gene Name
mitochondrial ribosomal protein S5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001321995.1 Intron NP_001308924.1
NM_001321996.1 Intron NP_001308925.1
NM_001321997.1 Intron NP_001308926.1
NM_031902.4 Intron NP_114108.1

View Full Product Details