Product Details

SNP ID
rs143245353
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:219283431 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGTCTCCCACAGAATCATGGAGAA[C/T]GGGCAGGAGCGGGTGGAAGTGGAGG
Phenotype
MIM: 604139 MIM: 601773
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
DNAJB2 PubMed Links

Gene Details

Gene
DNAJB2
Gene Name
DnaJ heat shock protein family (Hsp40) member B2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001039550.1 798 Silent Mutation AAC,AAT N187N NP_001034639.1
NM_006736.5 798 Silent Mutation AAC,AAT N187N NP_006727.2
Gene
PTPRN
Gene Name
protein tyrosine phosphatase, receptor type N
There are no transcripts associated with this gene.

View Full Product Details