Product Details

SNP ID
rs143352388
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:241851056 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGAACACTGGTGGCCAAGGAAGCC[A/G]GTCAGAGGGGCCAAGAGCAGTGTCC
Phenotype
MIM: 600244
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
PDCD1 PubMed Links

Gene Details

Gene
PDCD1
Gene Name
programmed cell death 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005018.2 936 UTR 3 NP_005009.2
XM_006712573.2 936 Intron XP_006712636.1
XM_017004293.1 936 UTR 3 XP_016859782.1

View Full Product Details