Product Details

SNP ID
rs143494796
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:3499049 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGGTTGTACGCTGTCCACACCGGTT[C/T]TCCCACAAACAGCCGCATGGCCTTC
Phenotype
MIM: 613400
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ADI1 PubMed Links

Gene Details

Gene
ADI1
Gene Name
acireductone dioxygenase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001306077.1 3274 Missense Mutation AAA,GAA K146E NP_001293006.1
NM_018269.3 3274 Missense Mutation AAA,GAA K152E NP_060739.2

View Full Product Details