Product Details

SNP ID
rs143561900
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:9428804 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAGTAGGAAGATCATGTATTATTCT[C/T]GAGTTCAAAGGAAGAAAAATAATGG
Phenotype
MIM: 606029 MIM: 607153
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CPSF3 PubMed Links

Gene Details

Gene
CPSF3
Gene Name
cleavage and polyadenylation specific factor 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001321833.1 556 UTR 5 NP_001308762.1
NM_001321834.1 556 UTR 5 NP_001308763.1
NM_001321835.1 556 UTR 5 NP_001308764.1
NM_001321836.1 556 Silent Mutation CTC,CTT L34L NP_001308765.1
NM_016207.3 556 Silent Mutation CTC,CTT L30L NP_057291.1
Gene
ITGB1BP1
Gene Name
integrin subunit beta 1 binding protein 1
There are no transcripts associated with this gene.

View Full Product Details