Product Details

SNP ID
rs143659490
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:99291365 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGCCAATGGTTTGCATCATGGGTT[C/G]ATATTTCAGGAGGTATGGCATGTCT
Phenotype
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
LYG1 PubMed Links

Gene Details

Gene
LYG1
Gene Name
lysozyme g1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_174898.2 439 Missense Mutation CAA,GAA Q69E NP_777558.1
XM_005263869.2 439 Missense Mutation CAA,GAA Q69E XP_005263926.1
XM_005263870.3 439 Missense Mutation CAA,GAA Q69E XP_005263927.1
XM_005263871.4 439 Missense Mutation CAA,GAA Q69E XP_005263928.1
XM_011510580.2 439 Missense Mutation CAA,GAA Q69E XP_011508882.1
XM_017003311.1 439 Missense Mutation CAA,GAA Q69E XP_016858800.1
XM_017003312.1 439 Missense Mutation CAA,GAA Q69E XP_016858801.1
XM_017003313.1 439 Missense Mutation CAA,GAA Q69E XP_016858802.1

View Full Product Details