Product Details

SNP ID
rs143762187
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:135912365 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTGGTGACAGACATCAATGGTAATA[C/T]GTATGTCTATCAAAACTGCAAATTA
Phenotype
MIM: 603084
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
DARS PubMed Links

Gene Details

Gene
DARS
Gene Name
aspartyl-tRNA synthetase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001293312.1 Intron NP_001280241.1
NM_001349.3 Intron NP_001340.2
XM_017003489.1 Intron XP_016858978.1

View Full Product Details