Product Details

SNP ID
rs143957989
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:68158432 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCAGTCCCAGCTAACAGATACACAC[C/T]ATTGAAAGAAAACTGGATGAAGATA
Phenotype
MIM: 610729
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PNO1 PubMed Links

Gene Details

Gene
PNO1
Gene Name
partner of NOB1 homolog
There are no transcripts associated with this gene.

Gene
WDR92
Gene Name
WD repeat domain 92
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001256476.1 Intron NP_001243405.1
NM_138458.3 Intron NP_612467.1

View Full Product Details