Product Details
- SNP ID
-
rs143968577
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.2:10058870 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TGCTGTCCTCAGCGGCAGTACTCCC[C/G]CTCGATGCTCGCCATCAGCCCCTCT
- Phenotype
-
MIM: 603301
- Polymorphism
- C/G, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
CYS1
PubMed Links
Gene Details
- Gene
- CYS1
- Gene Name
- cystin 1
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001037160.2 |
648 |
Missense Mutation |
CGG,GGG |
R154G |
NP_001032237.1 |
- Gene
- KLF11
- Gene Name
- Kruppel like factor 11
There are no transcripts associated with this gene.
View Full Product Details