Product Details

SNP ID
rs144164793
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:97648061 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAAAGGCGAGGCCCAGCGATGCCCC[C/T]GCTGTGGAGCCCATTACAAGCTGGT
Phenotype
MIM: 605144 MIM: 123866
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ACTR1B PubMed Links

Gene Details

Gene
ACTR1B
Gene Name
ARP1 actin-related protein 1 homolog B, centractin beta
There are no transcripts associated with this gene.

Gene
COX5B
Gene Name
cytochrome c oxidase subunit 5B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001862.2 Intron NP_001853.2

View Full Product Details