Product Details
- SNP ID
-
rs144175890
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.2:191836615 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GAGGAGAGGTCCACGGTGTGCAGGG[C/T]TTCCTCCAGGGATTTGTTTTCATCC
- Phenotype
-
MIM: 606728
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
SDPR
PubMed Links
Gene Details
- Gene
- SDPR
- Gene Name
- serum deprivation response
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_004657.5 |
941 |
Missense Mutation |
ACC,GCC |
T196A |
NP_004648.1 |
View Full Product Details