Product Details

SNP ID
rs144519975
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:161993299 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTGAGGTGCTAAGGTAAAGAGAAA[C/G]ATTGTTTTATGAAGTGGCTCATGTG
Phenotype
MIM: 102720 MIM: 605556
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
DPP4 PubMed Links

Gene Details

Gene
DPP4
Gene Name
dipeptidyl peptidase 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001935.3 2846 Missense Mutation TCT,TGT S762C NP_001926.2
XM_005246371.2 2846 Missense Mutation TCT,TGT S761C XP_005246428.1
Gene
SLC4A10
Gene Name
solute carrier family 4 member 10
There are no transcripts associated with this gene.

View Full Product Details