Product Details

SNP ID
rs144580593
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:130139618 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTGCAGCCTCTTCTGCCTCTCGGC[A/G]AAGTTGTTCTTCGGGGTCTGCTTCA
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CCDC74B PubMed Links

Gene Details

Gene
CCDC74B
Gene Name
coiled-coil domain containing 74B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001258307.1 Intron NP_001245236.1
NM_207310.2 Intron NP_997193.1
XM_005263840.2 Intron XP_005263897.1
XM_005263842.2 Intron XP_005263899.1
XM_006712833.2 Intron XP_006712896.1
XM_006712834.3 Intron XP_006712897.1
XM_006712835.2 Intron XP_006712898.1
XM_006712837.2 Intron XP_006712900.1
XM_011512142.2 Intron XP_011510444.1
XM_011512147.1 Intron XP_011510449.1
Gene
MED15P9
Gene Name
mediator complex subunit 15 pseudogene 9
There are no transcripts associated with this gene.

View Full Product Details