Product Details

SNP ID
rs144664229
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:207763213 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATATATTATGTATATATACAACCCA[C/T]AAAACATACATACTATACATAGTTT
Phenotype
MIM: 601723
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CCNYL1 PubMed Links

Gene Details

Gene
CCNYL1
Gene Name
cyclin Y like 1
There are no transcripts associated with this gene.

Gene
FZD5
Gene Name
frizzled class receptor 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003468.3 5937 UTR 3 NP_003459.2
Gene
MIR4775
Gene Name
microRNA 4775
There are no transcripts associated with this gene.

View Full Product Details