Product Details

SNP ID
rs144822809
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:30234459 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGAGGTGATGATGAACACCCAGCC[A/C]ATGGAGGAGATCGGCCTCAGCCCCC
Phenotype
MIM: 611763
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
LBH PubMed Links

Gene Details

Gene
LBH
Gene Name
limb bud and heart development
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_030915.3 289 Silent Mutation CCA,CCC P27P NP_112177.2
XM_017005059.1 289 Silent Mutation CCA,CCC P144P XP_016860548.1

View Full Product Details