Product Details

SNP ID
rs145077544
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:43775099 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTGTATAGGCCATGCCCTAAACTAC[A/C]TATATATTTTAACAATTATGACAAT
Phenotype
MIM: 612723
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
DYNC2LI1 PubMed Links

Gene Details

Gene
DYNC2LI1
Gene Name
dynein cytoplasmic 2 light intermediate chain 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001193464.1 Intron NP_001180393.1
NM_015522.3 Intron NP_056337.1
NM_016008.3 Intron NP_057092.2
XM_005264364.4 Intron XP_005264421.1
XM_005264365.4 Intron XP_005264422.1
Gene
PLEKHH2
Gene Name
pleckstrin homology, MyTH4 and FERM domain containing H2
There are no transcripts associated with this gene.

View Full Product Details