Product Details

SNP ID
rs145082926
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:105339681 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TACCATTGCCTCAGAGGGATTTGCC[A/G]AAGAATAGATGGGGGAAAATGATGG
Phenotype
MIM: 606237
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
C2orf49 PubMed Links

Gene Details

Gene
C2orf49
Gene Name
chromosome 2 open reading frame 49
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001286537.1 529 Silent Mutation CCA,CCG P66P NP_001273466.1
NM_024093.2 529 Silent Mutation CCA,CCG P66P NP_076998.1
XM_017004892.1 529 Silent Mutation CCA,CCG P104P XP_016860381.1
Gene
LOC100506473
Gene Name
uncharacterized LOC100506473
There are no transcripts associated with this gene.

Gene
TGFBRAP1
Gene Name
transforming growth factor beta receptor associated protein 1
There are no transcripts associated with this gene.

View Full Product Details