Product Details

SNP ID
rs145137776
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:1790899 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCTTAAATTATCTTTTAGCTTAAAT[C/T]GAAAACACACAAATTCTGTTGATAA
Phenotype
MIM: 613084
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MYT1L PubMed Links

Gene Details

Gene
MYT1L
Gene Name
myelin transcription factor 1 like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001303052.1 5637 UTR 3 NP_001289981.1
NM_015025.3 5637 UTR 3 NP_055840.2
XM_011510318.1 5637 UTR 3 XP_011508620.1
XM_011510319.1 5637 UTR 3 XP_011508621.1
XM_011510320.2 5637 UTR 3 XP_011508622.1
XM_011510321.2 5637 UTR 3 XP_011508623.1
XM_011510322.2 5637 UTR 3 XP_011508624.1
XM_011510323.1 5637 UTR 3 XP_011508625.1
XM_011510324.1 5637 UTR 3 XP_011508626.1
XM_011510325.1 5637 UTR 3 XP_011508627.1
XM_011510326.1 5637 UTR 3 XP_011508628.1
XM_011510327.1 5637 UTR 3 XP_011508629.1
XM_011510328.2 5637 UTR 3 XP_011508630.1
XM_011510331.2 5637 Intron XP_011508633.1
XM_011510332.2 5637 Intron XP_011508634.1
XM_017003604.1 5637 UTR 3 XP_016859093.1
XM_017003605.1 5637 UTR 3 XP_016859094.1
XM_017003606.1 5637 UTR 3 XP_016859095.1
XM_017003607.1 5637 UTR 3 XP_016859096.1
XM_017003608.1 5637 Intron XP_016859097.1
XM_017003609.1 5637 UTR 3 XP_016859098.1
XM_017003610.1 5637 UTR 3 XP_016859099.1
XM_017003611.1 5637 UTR 3 XP_016859100.1
XM_017003612.1 5637 UTR 3 XP_016859101.1
XM_017003613.1 5637 UTR 3 XP_016859102.1
XM_017003614.1 5637 UTR 3 XP_016859103.1
XM_017003615.1 5637 UTR 3 XP_016859104.1
XM_017003616.1 5637 UTR 3 XP_016859105.1
XM_017003617.1 5637 UTR 3 XP_016859106.1
XM_017003618.1 5637 UTR 3 XP_016859107.1
XM_017003619.1 5637 UTR 3 XP_016859108.1
XM_017003620.1 5637 UTR 3 XP_016859109.1
XM_017003621.1 5637 Intron XP_016859110.1
XM_017003622.1 5637 Intron XP_016859111.1

View Full Product Details