Product Details

SNP ID
rs145203305
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:33583948 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TAAAGCAAACTAACTTGTATTTGCT[C/G]AGCCAGTGGTATTAACTCATGCATA
Phenotype
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
FAM98A PubMed Links

Gene Details

Gene
FAM98A
Gene Name
family with sequence similarity 98 member A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001304538.1 2526 UTR 3 NP_001291467.1
NM_015475.4 2526 UTR 3 NP_056290.3

View Full Product Details