Product Details

SNP ID
rs145273962
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:38984515 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTAAAACTTACTTACAAAGTTCTGC[C/T]TAAAAGATACCCATCCTATTCTAAC
Phenotype
MIM: 182530
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ARHGEF33 PubMed Links

Gene Details

Gene
ARHGEF33
Gene Name
Rho guanine nucleotide exchange factor 33
There are no transcripts associated with this gene.

Gene
SOS1
Gene Name
SOS Ras/Rac guanine nucleotide exchange factor 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005633.3 5520 UTR 3 NP_005624.2
XM_005264515.4 5520 UTR 3 XP_005264572.1
XM_011533062.2 5520 UTR 3 XP_011531364.1
XM_011533064.2 5520 UTR 3 XP_011531366.1
XM_011533066.2 5520 UTR 3 XP_011531368.1

View Full Product Details