Product Details

SNP ID
rs145331226
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:241704702 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTCAGAGGAACTTCCAGCTGATGCG[A/G]GAGCTGGACCAGAGGACGGAAGGTG
Phenotype
MIM: 608525
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ING5 PubMed Links

Gene Details

Gene
ING5
Gene Name
inhibitor of growth family member 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
XM_005247047.2 1180 Silent Mutation CGA,CGG R29R XP_005247104.1
XM_017005097.1 1180 Silent Mutation CGA,CGG R89R XP_016860586.1
XM_017005098.1 1180 Silent Mutation CGA,CGG R89R XP_016860587.1
XM_017005099.1 1180 Silent Mutation CGA,CGG R89R XP_016860588.1
XM_017005100.1 1180 Silent Mutation CGA,CGG R31R XP_016860589.1
XM_017005101.1 1180 Silent Mutation CGA,CGG R31R XP_016860590.1
XM_017005102.1 1180 Silent Mutation CGA,CGG R29R XP_016860591.1

View Full Product Details