Product Details

SNP ID
rs145550482
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:96129832 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTGCCATGCCACTCACCTCTCCCA[A/C]GGGGCCTGGGGCCACTTGGGCTGCA
Phenotype
MIM: 608860
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
ASTL PubMed Links

Gene Details

Gene
ASTL
Gene Name
astacin-like metallo-endopeptidase (M12 family)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001002036.3 1681 Missense Mutation NP_001002036.3
XM_011511205.2 1681 Missense Mutation XP_011509507.1
XM_011511206.2 1681 Missense Mutation XP_011509508.1
XM_011511207.2 1681 Missense Mutation XP_011509509.1
XM_011511208.2 1681 Intron XP_011509510.1

View Full Product Details