Product Details

SNP ID
rs145686945
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:27443455 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCTCATGTCTGCAGGGGAGGCTGCT[C/G]TATCTGGTTACTGCTGTGTGGCTGC
Phenotype
MIM: 607386 MIM: 606010
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
IFT172 PubMed Links

Gene Details

Gene
IFT172
Gene Name
intraflagellar transport 172
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015662.2 585 Intron NP_056477.1
XM_005264254.1 585 Intron XP_005264311.1
XM_006711986.3 585 Intron XP_006712049.1
XM_006711987.1 585 Intron XP_006712050.1
XM_011532757.2 585 Intron XP_011531059.1
XM_011532758.1 585 Intron XP_011531060.1
XM_011532759.2 585 Intron XP_011531061.1
XM_011532760.2 585 Intron XP_011531062.1
XM_017003790.1 585 Intron XP_016859279.1
XM_017003791.1 585 Intron XP_016859280.1
XM_017003792.1 585 Intron XP_016859281.1
XM_017003793.1 585 Intron XP_016859282.1
XM_017003794.1 585 Intron XP_016859283.1
XM_017003795.1 585 Intron XP_016859284.1
Gene
KRTCAP3
Gene Name
keratinocyte associated protein 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001168364.1 585 Missense Mutation CTA,GTA L180V NP_001161836.1
NM_001321325.1 585 Missense Mutation CTA,GTA L180V NP_001308254.1
NM_173853.3 585 Missense Mutation CTA,GTA L180V NP_776252.2
Gene
NRBP1
Gene Name
nuclear receptor binding protein 1
There are no transcripts associated with this gene.

View Full Product Details