Product Details

SNP ID
rs145773754
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:119440208 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAAGGAGGCCACGGGGTGCAGTGGG[A/G]ACTCACGGAGGTGCCATTGCTGCCA
Phenotype
MIM: 182098
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SCTR PubMed Links

Gene Details

Gene
SCTR
Gene Name
secretin receptor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002980.2 831 Missense Mutation TCC,TTC S411F NP_002971.2
XM_005263730.4 831 Missense Mutation TCC,TTC S233F XP_005263787.1
XM_011511621.2 831 Missense Mutation TCC,TTC S416F XP_011509923.1
XM_017004670.1 831 Missense Mutation TCC,TTC S406F XP_016860159.1
XM_017004671.1 831 Missense Mutation TCC,TTC S380F XP_016860160.1
XM_017004672.1 831 Missense Mutation TCC,TTC S348F XP_016860161.1
XM_017004673.1 831 Missense Mutation TCC,TTC S238F XP_016860162.1
Gene
TMEM37
Gene Name
transmembrane protein 37
There are no transcripts associated with this gene.

View Full Product Details