Product Details
- SNP ID
-
rs146035954
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.2:68946379 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TTCTCAATGGGTGACCCAAGCAGGA[A/G]CCAATCCACGTCTTTGATCAGAGAC
- Phenotype
-
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
GKN2
PubMed Links
Gene Details
- Gene
- GKN2
- Gene Name
- gastrokine 2
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_182536.2 |
506 |
Missense Mutation |
CTC,TTC |
L133F |
NP_872342.2 |
View Full Product Details