Product Details

SNP ID
rs146151167
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:202880879 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCACTTTCATGCCCCAACATGGGAA[A/G]TGGTAGGTGAATATCTGTAGGAATA
Phenotype
MIM: 616620
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ICA1L PubMed Links

Gene Details

Gene
ICA1L
Gene Name
islet cell autoantigen 1 like
There are no transcripts associated with this gene.

Gene
WDR12
Gene Name
WD repeat domain 12
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_018256.3 1880 Missense Mutation ACT,ATT T418I NP_060726.3
XM_011511469.2 1880 Missense Mutation ACT,ATT T280I XP_011509771.1

View Full Product Details