Product Details

SNP ID
rs146273697
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:86214554 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGTCTGTAAGTCAGCAAATTAAAAA[C/T]ATTCAGTTGTATCCTCTAGACAGAA
Phenotype
MIM: 611841 MIM: 609139
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MRPL35 PubMed Links

Gene Details

Gene
MRPL35
Gene Name
mitochondrial ribosomal protein L35
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_016622.3 3402 Intron NP_057706.2
NM_145644.2 3402 Intron NP_663619.1
XM_005264361.2 3402 Intron XP_005264418.1
Gene
REEP1
Gene Name
receptor accessory protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001164730.1 3402 UTR 3 NP_001158202.1
NM_001164731.1 3402 UTR 3 NP_001158203.1
NM_001164732.1 3402 UTR 3 NP_001158204.1
NM_022912.2 3402 UTR 3 NP_075063.1
XM_005264502.1 3402 UTR 3 XP_005264559.1
XM_005264504.1 3402 UTR 3 XP_005264561.1
XM_011533044.1 3402 UTR 3 XP_011531346.1
XM_011533045.2 3402 UTR 3 XP_011531347.1
XM_017004725.1 3402 UTR 3 XP_016860214.1
XM_017004726.1 3402 Intron XP_016860215.1
XM_017004727.1 3402 UTR 3 XP_016860216.1

View Full Product Details