Product Details

SNP ID
rs146329964
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:240126676 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCCCTCCAGTGAGTAGAAACTGATA[G/T]TGTGCTGTCTTCGCTGACCTCTCGC
Phenotype
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
COPS9 PubMed Links

Gene Details

Gene
COPS9
Gene Name
COP9 signalosome subunit 9
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001163424.1 1071 Intron NP_001156896.1
NM_138336.1 1071 Missense Mutation ATA,CTA I216L NP_612209.1
XM_017003411.1 1071 Missense Mutation ATA,CTA I206L XP_016858900.1

View Full Product Details