Product Details

SNP ID
rs146355232
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:112485960 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TACAGTTGGTGAATTACTACAGGGG[C/T]GCTGACAAACTGTGTCGCAAAGCTT
Phenotype
MIM: 608291
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
TTL PubMed Links

Gene Details

Gene
TTL
Gene Name
tubulin tyrosine ligase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_153712.4 419 Silent Mutation GGC,GGT G67G NP_714923.1
XM_005263599.3 419 Silent Mutation GGC,GGT G67G XP_005263656.1
XM_011510665.2 419 Silent Mutation GGC,GGT G67G XP_011508967.1

View Full Product Details