Product Details

SNP ID
rs146528502
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:237566607 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCTGAGGGCCTGGCTCCTGTGCCTG[A/C]TGATGCTGGGCCTGGCCCTGCGGGG
Phenotype
MIM: 602663 MIM: 602206
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
PRLH PubMed Links

Gene Details

Gene
PRLH
Gene Name
prolactin releasing hormone
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015893.1 34 Missense Mutation ATG,CTG M12L NP_056977.1
Gene
RAB17
Gene Name
RAB17, member RAS oncogene family
There are no transcripts associated with this gene.

View Full Product Details