Product Details

SNP ID
rs146619645
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:236198196 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AATATGACAAACTAAAAAAAATTCA[C/T]ATTCCTCTGGCTAGAGGTAACTACT
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ASB18 PubMed Links

Gene Details

Gene
ASB18
Gene Name
ankyrin repeat and SOCS box containing 18
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_212556.2 Intron NP_997721.2

View Full Product Details