Product Details
- SNP ID
-
rs146684990
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.2:189447203 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TATAATAGACACAGAGCAAATACTT[C/T]TTACTTGGAAGATACAAGAAGATTT
- Phenotype
-
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
WDR75
PubMed Links
Gene Details
- Gene
- WDR75
- Gene Name
- WD repeat domain 75
View Full Product Details