Product Details

SNP ID
rs146696981
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:201186003 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGAGGATCTGCTGAGTGAGGAAGAC[A/C]CTTTCTTCCTGGCAGAACTCCTCTA
Phenotype
MIM: 601762
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
CASP10 PubMed Links

Gene Details

Gene
CASP10
Gene Name
caspase 10
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001206524.1 644 Missense Mutation ACT,CCT T76P NP_001193453.1
NM_001206542.1 644 Missense Mutation ACT,CCT T76P NP_001193471.1
NM_001230.4 644 Missense Mutation ACT,CCT T76P NP_001221.2
NM_001306083.1 644 Missense Mutation ACT,CCT T76P NP_001293012.1
NM_032974.4 644 Missense Mutation ACT,CCT T76P NP_116756.2
NM_032976.3 644 Missense Mutation ACT,CCT T76P NP_116758.1
NM_032977.3 644 Missense Mutation ACT,CCT T76P NP_116759.2
XM_005246907.2 644 Missense Mutation ACT,CCT T76P XP_005246964.1
XM_006712796.3 644 UTR 5 XP_006712859.1
XM_011511990.2 644 Missense Mutation ACT,CCT T76P XP_011510292.1

View Full Product Details