Product Details
- SNP ID
-
rs146697305
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.2:3499033 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AAAATGGTCAGCGGGCCGGTTGTAC[A/G]CTGTCCACACCGGTTCTCCCACAAA
- Phenotype
-
MIM: 613400
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
ADI1
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs10208494] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- ADI1
- Gene Name
- acireductone dioxygenase 1
View Full Product Details