Product Details

SNP ID
rs146705876
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:69960866 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATGGCTTCCTCGGCACTCGCATTGG[C/T]CACTAGGAACTGTGCCTTCAGCTTC
Phenotype
MIM: 611765
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ASPRV1 PubMed Links

Gene Details

Gene
ASPRV1
Gene Name
aspartic peptidase, retroviral-like 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_152792.2 1400 Missense Mutation ACC,GCC T275A NP_690005.2
Gene
PCBP1-AS1
Gene Name
PCBP1 antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details