Product Details

SNP ID
rs146713767
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:171522589 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGCGCTTCCTGGCGCTGCTGGGGT[C/T]GGCACTGCTCGTCGGCTTCCTGTCG
Phenotype
MIM: 605745
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CYBRD1 PubMed Links

Gene Details

Gene
CYBRD1
Gene Name
cytochrome b reductase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001127383.1 234 Missense Mutation TCG,TTG S15L NP_001120855.1
NM_001256909.1 234 Intron NP_001243838.1
NM_024843.3 234 Missense Mutation TCG,TTG S15L NP_079119.3

View Full Product Details