Product Details

SNP ID
rs146765948
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:61872125 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGCAGTTTTTTCTCCCTGGGCATGC[C/T]GGTTTCTTAGTTCTGTTACTGTAGA
Phenotype
MIM: 605142
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CCT4 PubMed Links

Gene Details

Gene
CCT4
Gene Name
chaperonin containing TCP1 subunit 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001256721.1 1612 Missense Mutation CAG,CGG Q453R NP_001243650.1
NM_006430.3 1612 Missense Mutation CAG,CGG Q483R NP_006421.2

View Full Product Details