Product Details

SNP ID
rs146784144
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:39254733 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGTAGGAAAAAATTCACCGAAAATA[C/T]TTTTGTTATAGTTTTTTTGTATTTC
Phenotype
MIM: 604921
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CDKL4 PubMed Links

Gene Details

Gene
CDKL4
Gene Name
cyclin dependent kinase like 4
There are no transcripts associated with this gene.

Gene
MAP4K3
Gene Name
mitogen-activated protein kinase kinase kinase kinase 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001270425.1 Intron NP_001257354.1
NM_003618.3 Intron NP_003609.2

View Full Product Details