Product Details

SNP ID
rs146938630
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:178483571 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTGCCCAATATTTGTTTTAAAGAG[A/C]TATTGAATTGCAGTACTGCAGGCTT
Phenotype
MIM: 607062 MIM: 607774
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
FKBP7 PubMed Links

Gene Details

Gene
FKBP7
Gene Name
FK506 binding protein 7
There are no transcripts associated with this gene.

Gene
PLEKHA3
Gene Name
pleckstrin homology domain containing A3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_019091.3 Intron NP_061964.3
XM_011511689.2 Intron XP_011509991.1

View Full Product Details