Product Details

SNP ID
rs147007578
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:39258351 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAATTATAAAACTTTGACTTACCTA[C/T]TGATTCAATCTGGAAATCAAAGGTG
Phenotype
MIM: 604921
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MAP4K3 PubMed Links

Gene Details

Gene
MAP4K3
Gene Name
mitogen-activated protein kinase kinase kinase kinase 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001270425.1 2792 Missense Mutation ATA,GTA I802V NP_001257354.1
NM_003618.3 2792 Missense Mutation ATA,GTA I823V NP_003609.2

View Full Product Details