Product Details

SNP ID
rs147267731
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.2:96124191 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCACTGGGGTCGGGGCTCCTGGATT[C/T]CGCCGACAGTGCCTCCAAAAGCCGC
Phenotype
MIM: 104260 MIM: 608860
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
ADRA2B PubMed Links

Gene Details

Gene
ADRA2B
Gene Name
adrenoceptor alpha 2B
There are no transcripts associated with this gene.

Gene
ASTL
Gene Name
astacin-like metallo-endopeptidase (M12 family)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001002036.3 1770 Missense Mutation AAA,GAA K319E NP_001002036.3
XM_011511205.2 1770 Missense Mutation AAA,GAA K324E XP_011509507.1
XM_011511206.2 1770 Missense Mutation AAA,GAA K320E XP_011509508.1
XM_011511207.2 1770 Missense Mutation AAA,GAA K306E XP_011509509.1
XM_011511208.2 1770 UTR 3 XP_011509510.1

View Full Product Details