Product Details

SNP ID
rs147343205
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:218610685 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAGGCCCTTTCCAAGAGCAGAGTCA[C/T]TGTGAGATCCTTGAGCTGGTTGTAT
Phenotype
MIM: 605939
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PLCD4 PubMed Links

Gene Details

Gene
PLCD4
Gene Name
phospholipase C delta 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_032726.3 Intron NP_116115.1
XM_005246913.2 Intron XP_005246970.1
XM_011512012.1 Intron XP_011510314.1
XM_017005114.1 Intron XP_016860603.1
XM_017005115.1 Intron XP_016860604.1
XM_017005116.1 Intron XP_016860605.1
XM_017005117.1 Intron XP_016860606.1
XM_017005118.1 Intron XP_016860607.1
XM_017005119.1 Intron XP_016860608.1
XM_017005120.1 Intron XP_016860609.1

View Full Product Details