Product Details

SNP ID
rs148093004
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:241153811 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACTCATTGATGGCGTTTGCAAAAAG[G/T]ATGTGGTTTTTTCCAATTCCACTGA
Phenotype
MIM: 607505 MIM: 602877
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
PASK PubMed Links

Gene Details

Gene
PASK
Gene Name
PAS domain containing serine/threonine kinase
There are no transcripts associated with this gene.

Gene
PPP1R7
Gene Name
protein phosphatase 1 regulatory subunit 7
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001282409.1 Intron NP_001269338.1
NM_001282410.1 Intron NP_001269339.1
NM_001282411.1 Intron NP_001269340.1
NM_001282412.1 Intron NP_001269341.1
NM_001282413.1 Intron NP_001269342.1
NM_001282414.1 Intron NP_001269343.1
NM_002712.2 Intron NP_002703.1
XM_011511388.1 Intron XP_011509690.1

View Full Product Details