Product Details

SNP ID
rs148102142
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:178118094 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGAGTATTGCAGCATTATTAAGAAT[A/T]AAGTGACTGGAGAAAGTAAAGGTTT
Phenotype
MIM: 604961 MIM: 608888
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
PDE11A PubMed Links

Gene Details

Gene
PDE11A
Gene Name
phosphodiesterase 11A
There are no transcripts associated with this gene.

Gene
RBM45
Gene Name
RNA binding motif protein 45
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_152945.3 1030 Nonsense Mutation AAA,TAA K155* NP_694453.2
XM_005246287.4 1030 Nonsense Mutation AAA,TAA K155* XP_005246344.1
XM_017003320.1 1030 Nonsense Mutation AAA,TAA K298* XP_016858809.1
XM_017003321.1 1030 Nonsense Mutation AAA,TAA K298* XP_016858810.1
XM_017003322.1 1030 Nonsense Mutation AAA,TAA K155* XP_016858811.1

View Full Product Details