Product Details
- SNP ID
-
rs148102267
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.2:241108289 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TCCCAGAGACCACATCTCCAGCTCC[A/G]GCCCTCTGTAGCTGTGAGGAGGAGG
- Phenotype
-
MIM: 615393
MIM: 607505
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
MTERF4
PubMed Links
Gene Details
- Gene
- MTERF4
- Gene Name
- mitochondrial transcription termination factor 4
There are no transcripts associated with this gene.
- Gene
- PASK
- Gene Name
- PAS domain containing serine/threonine kinase
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001252119.1 |
3840 |
Missense Mutation |
CCG,CTG |
P1189L |
NP_001239048.1 |
NM_001252120.1 |
3840 |
Missense Mutation |
CCG,CTG |
P1182L |
NP_001239049.1 |
NM_001252122.1 |
3840 |
Missense Mutation |
CCG,CTG |
P1147L |
NP_001239051.1 |
NM_001252124.1 |
3840 |
Intron |
|
|
NP_001239053.1 |
NM_015148.3 |
3840 |
Missense Mutation |
CCG,CTG |
P1182L |
NP_055963.2 |
XM_005246991.1 |
3840 |
Missense Mutation |
CCG,CTG |
P966L |
XP_005247048.1 |
XM_011510826.1 |
3840 |
Missense Mutation |
CCG,CTG |
P1214L |
XP_011509128.1 |
XM_011510828.1 |
3840 |
Missense Mutation |
CCG,CTG |
P1189L |
XP_011509130.1 |
XM_011510829.1 |
3840 |
Missense Mutation |
CCG,CTG |
P1189L |
XP_011509131.1 |
XM_011510830.1 |
3840 |
Missense Mutation |
CCG,CTG |
P1189L |
XP_011509132.1 |
XM_011510831.2 |
3840 |
Missense Mutation |
CCG,CTG |
P1003L |
XP_011509133.1 |
XM_011510832.2 |
3840 |
Missense Mutation |
CCG,CTG |
P996L |
XP_011509134.1 |
XM_011510833.1 |
3840 |
Missense Mutation |
CCG,CTG |
P973L |
XP_011509135.1 |
XM_011510834.1 |
3840 |
Missense Mutation |
CCG,CTG |
P803L |
XP_011509136.1 |
XM_011510835.1 |
3840 |
Missense Mutation |
CCG,CTG |
P732L |
XP_011509137.1 |
XM_017003634.1 |
3840 |
Missense Mutation |
CCG,CTG |
P1273L |
XP_016859123.1 |
XM_017003635.1 |
3840 |
Missense Mutation |
CCG,CTG |
P1182L |
XP_016859124.1 |
XM_017003636.1 |
3840 |
Missense Mutation |
CCG,CTG |
P1182L |
XP_016859125.1 |
View Full Product Details