Product Details

SNP ID
rs148115152
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:96124069 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAGCTAGGGTCTGAGGCTGCCTTGC[C/T]GAGGCCTCTGCACTGAGCTTTTTCA
Phenotype
MIM: 104260 MIM: 608860
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ADRA2B PubMed Links

Gene Details

Gene
ADRA2B
Gene Name
adrenoceptor alpha 2B
There are no transcripts associated with this gene.

Gene
ASTL
Gene Name
astacin-like metallo-endopeptidase (M12 family)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001002036.3 1892 Silent Mutation TCA,TCG S359S NP_001002036.3
XM_011511205.2 1892 Silent Mutation TCA,TCG S364S XP_011509507.1
XM_011511206.2 1892 Silent Mutation TCA,TCG S360S XP_011509508.1
XM_011511207.2 1892 Silent Mutation TCA,TCG S346S XP_011509509.1
XM_011511208.2 1892 Intron XP_011509510.1

View Full Product Details