Product Details

SNP ID
rs148530634
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:197501208 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAGGAAAAGTATTGCAAGCAACAGT[A/G]GTCGCTGTTGGATCGGGTTCTAAAG
Phenotype
MIM: 118190 MIM: 600141
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
HSPD1 PubMed Links

Gene Details

Gene
HSPD1
Gene Name
heat shock protein family D (Hsp60) member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002156.4 578 Intron NP_002147.2
NM_199440.1 578 Intron NP_955472.1
Gene
HSPE1
Gene Name
heat shock protein family E (Hsp10) member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002157.2 578 Silent Mutation GTA,GTG V46V NP_002148.1
Gene
HSPE1-MOB4
Gene Name
HSPE1-MOB4 readthrough
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001202485.1 578 Silent Mutation GTA,GTG V46V NP_001189414.1

View Full Product Details