Product Details
- SNP ID
-
rs148732839
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.2:70809782 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TTGAATGGTGTCCCATCTGTCCAGC[A/G]CCAGGAGCCCTCTGTGCCCCTGTCA
- Phenotype
-
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
CLEC4F
PubMed Links
Gene Details
- Gene
- CLEC4F
- Gene Name
- C-type lectin domain family 4 member F
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001258027.1 |
1692 |
Missense Mutation |
CGC,TGC |
R539C |
NP_001244956.1 |
NM_001321308.1 |
1692 |
Missense Mutation |
CGC,TGC |
R539C |
NP_001308237.1 |
NM_173535.2 |
1692 |
Missense Mutation |
CGC,TGC |
R539C |
NP_775806.2 |
XM_011532635.2 |
1692 |
Missense Mutation |
CGC,TGC |
R649C |
XP_011530937.1 |
XM_011532637.2 |
1692 |
Missense Mutation |
CGC,TGC |
R649C |
XP_011530939.1 |
XM_011532638.2 |
1692 |
Missense Mutation |
CGC,TGC |
R649C |
XP_011530940.1 |
XM_011532639.2 |
1692 |
Missense Mutation |
CGC,TGC |
R649C |
XP_011530941.1 |
XM_011532640.2 |
1692 |
Missense Mutation |
CGC,TGC |
R649C |
XP_011530942.1 |
XM_011532641.2 |
1692 |
UTR 3 |
|
|
XP_011530943.1 |
XM_011532642.2 |
1692 |
Missense Mutation |
CGC,TGC |
R509C |
XP_011530944.1 |
XM_011532643.1 |
1692 |
Missense Mutation |
CGC,TGC |
R478C |
XP_011530945.1 |
XM_017003519.1 |
1692 |
Missense Mutation |
CGC,TGC |
R649C |
XP_016859008.1 |
View Full Product Details