Product Details

SNP ID
rs148787671
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:96500763 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGCCGCTCTCCTCCCGCAGCACTC[G/T]CAGCGCCACACGCTCGCCCAGGCGC
Phenotype
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
NEURL3 PubMed Links

Gene Details

Gene
NEURL3
Gene Name
neuralized E3 ubiquitin protein ligase 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001285485.1 601 Silent Mutation AGA,CGA R64R NP_001272414.1
NM_001285486.1 601 UTR 5 NP_001272415.1
XM_011512178.2 601 Silent Mutation AGA,CGA R64R XP_011510480.1
XM_011512179.2 601 Silent Mutation AGA,CGA R64R XP_011510481.1

View Full Product Details