Product Details

SNP ID
rs148821869
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:68464418 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAAAAAGACTGTAGTAGGTGGTTGG[C/T]TAGTGAATGTGTTTAAAGCAAGAGA
Phenotype
MIM: 611035
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
APLF PubMed Links

Gene Details

Gene
APLF
Gene Name
aprataxin and PNKP like factor
There are no transcripts associated with this gene.

Gene
FBXO48
Gene Name
F-box protein 48
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001024680.2 Intron NP_001019851.1
XM_005264407.2 Intron XP_005264464.1
XM_017004437.1 Intron XP_016859926.1

View Full Product Details